missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Indolethylamine N-methyltransferase Protein

A DNA sequence encoding the Homo sapiens (Human) Indolethylamine N-methyltransferase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6233-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16091790

  • 2248.90 € / 1mg

Estimated Shipment: 12-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_001186148.1
Indolethylamine N-methyltransferase Protein
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
11185
Greater than 90% as determined by SDS-PAGE.
Research Use Only
INMT
Recombinant Protein
E. coli
MKGGFTGGDE YQKHFLPRDY LATYYSFDGS PSPEAEMLKF NLECLHKTFG PGGLQGDTLI DIGSGPTIYQ VLAACDSFQD ITLSDFTDRN REELEKWLKK EPGAYDWTPA VKFACELEGN SGRWEEKEEK LRAAVKRVLK CDVHLGNPLA PAVLPLADCV LTLLAMECAC CSLDAYRAAL CNLASLLKPG GHLVTTVTLR LPSYMVGKRE FSCVALEKEE VEQAVLDAGF DIEQLLHSPQ SYSVTNAANN GVCFIVARKK PGP
Videos
SDS
Documents
Certificates
Promotions

Promotions