missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Indolethylamine N-methyltransferase Protein

A DNA sequence encoding the Homo sapiens (Human) Indolethylamine N-methyltransferase, was expressed in the hosts and tags indicated.

160.00 € - 2385.00 €

Specifications

Accession Number NP_001186148.1
Gene ID (Entrez) 11185
Name Indolethylamine N-methyltransferase Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16071790
View Documents Promotion Details
enQuireBio™
QP6233-EC-100UG
100 μg
530.00 €
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 12-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16091790
View Documents Promotion Details
enQuireBio™
QP6233-EC-1MG
1 mg
2385.00 €
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 12-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16081790
View Documents Promotion Details
enQuireBio™
QP6233-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16001800
View Documents Promotion Details
enQuireBio™
QP6233-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16011800
View Documents Promotion Details
enQuireBio™
QP6233-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16021800
View Documents Promotion Details
enQuireBio™
QP6233-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_001186148.1
Indolethylamine N-methyltransferase Protein
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
11185
Research Use Only
INMT
Recombinant Protein
E. coli
MKGGFTGGDE YQKHFLPRDY LATYYSFDGS PSPEAEMLKF NLECLHKTFG PGGLQGDTLI DIGSGPTIYQ VLAACDSFQD ITLSDFTDRN REELEKWLKK EPGAYDWTPA VKFACELEGN SGRWEEKEEK LRAAVKRVLK CDVHLGNPLA PAVLPLADCV LTLLAMECAC CSLDAYRAAL CNLASLLKPG GHLVTTVTLR LPSYMVGKRE FSCVALEKEE VEQAVLDAGF DIEQLLHSPQ SYSVTNAANN GVCFIVARKK PGP
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications