missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ILDR2 Protein
A DNA sequence encoding the Homo sapiens (Human) ILDR2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7756-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_955383.1 | |
ILDR2 Protein | |
Research Use Only | |
ILDR2 | |
Recombinant Protein | |
E. coli | |
MDRVLLRWIS LFWLTAMVEG LQVTVPDKKK VAMLFQPTVL RCHFSTSSHQ PAVVQWKFKS YCQDRMGESL GMSSTRAQSL SKRNLEWDPY LDCLDSRRTV RVVASKQGST VTLGDFYRGR EITIVHDADL QIGKLMWGDS GLYYCIITTP DDLEGKNEDS VELLVLGRTG LLADLLPSFA VEIMPE | |
Greater than 90% as determined by SDS-PAGE. |
387597 | |
1 mg | |
Not determined. | |
Extracellular Domain | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |