missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-4R alpha His-tag Avi-tag Protein
Biotinylated
Brand: R&D Systems™ AVI11165-025
This item is not returnable.
View return policy
Description
Measured by its binding ability in a functional ELISA. When Recombinant Human IL-4 (Catalog # 204-IL/CF ) is immobilized at 1 μg/mL (100 μL/well), Biotinylated Recombinant Human IL-4R alpha His-tag Avi-tag (Catalog # AVI11165) binds with an ED50 of 70.0-560 ng/mL.Specifications
P24394.1 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
M.W. Observed: 39-52 kDa, under reducing conditions., M.W. Theoretical: 26 kDa | |
Human embryonic kidney cell, HEK293-derived human IL-4R alpha protein Human IL-4R (Gly24-His232) (N-terminus) Accession # P24394.1 6-His tag Avi-tag (C-terminus) | |
<0.10 EU per 1 μg of the protein by the LAL method. | |
Biotin | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie® Blue Staining. |
Bioactivity | |
3566 | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. | |
CD124, CD124 antigen, IL-4 R alpha, IL-4 receptor subunit alpha, IL4R, IL4R alpha, IL-4R subunit alpha, IL-4RA, IL4RACD124, IL-4R-alpha, interleukin 4 receptor, interleukin-4 receptor alpha chain, interleukin-4 receptor subunit alpha | |
Reconstitute at 250 μg/mL in PBS. | |
IL-4R alpha |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction