missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL-1 beta / IL1B Protein

A DNA sequence encoding the Homo sapiens (Human) IL-1 beta / IL1B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6214-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16071630

Quantity Price
1 2248.90 € / 1mg
3 2115.77 € / 1mg
Estimated Shipment: 18-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_000567.1
IL-1 beta / IL1B Protein
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
3553
Greater than 90% as determined by SDS-PAGE.
Research Use Only
IL1B
Recombinant Protein
E. coli
APVRSLNCTL RDSQQKSLVM SGPYELKALH LQGQDMEQQV VFSMSFVQGE ESNDKIPVAL GLKEKNLYLS CVLKDDKPTL QLESVDPKNY PKKKMEKRFV FNKIEINNKL EFESAQFPNW YISTSQAENM PVFLGGTKGG QDITDFTMQF VSS
Videos
SDS
Documents
Certificates
Promotions

Promotions