missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IFN-alpha / IFNA1 / IFN Protein

A DNA sequence encoding the Homo sapiens (Human) IFN-alpha / IFNA1 / IFN, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7402-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16068680

  • 493.03 € / 100µg

Estimated Shipment: 10-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_008831.3
IFN-alpha / IFNA1 / IFN Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
3439
Greater than 90% as determined by SDS-PAGE.
Research Use Only
IFNA1
Recombinant Protein
E. coli
CDLPETHSLD NRRTLMLLAQ MSRISPSSCL MDRHDFGFPQ EEFDGNQFQK APAISVLHEL IQQIFNLFTT KDSSAAWDED LLDKFCTELY QQLNDLEACV MQEERVGETP LMNADSILAV KKYFRRITLY LTEKKYSPCA WEVVRAEIMR SLSLSTNLQE RLRRKE
Videos
SDS
Documents
Certificates
Promotions

Promotions