missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IDH3G, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) IDH3G, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6194-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004126.1 | |
IDH3G, mitochondrial Protein | |
Research Use Only | |
IDH3G | |
Recombinant Protein | |
E. coli | |
FSEQTIPPSA KYGGRHTVTM IPGDGIGPEL MLHVKSVFRH ACVPVDFEEV HVSSNADEED IRNAIMAIRR NRVALKGNIE TNHNLPPSHK SRNNILRTSL DLYANVIHCK SLPGVVTRHK DIDILIVREN TEGEYSSLEH ESVAGVVESL KIITKAKSLR IAEYAFKLAQ ESGRKKVTAV HKANIMKLGD GLFLQCCREV AARYPQITFE NMIVDNTTMQ LVSRPQQFDV MVMPNLYGNI VNNVCAGLVG GPGLVAGANY GHVYAVFETA TRNTGKSIAN KNIANPTATL LASCMMLDHL KLHSYATSIR KAVLASMDNE NMHTPDIGGQ GTTSEAIQDV IRHIRVINGR AVEA | |
Greater than 90% as determined by SDS-PAGE. |
3421 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |