missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IDH3G, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) IDH3G, mitochondrial, was expressed in the hosts and tags indicated.
160.00 € - 2385.00 €
Specifications
Accession Number | NP_004126.1 |
---|---|
Gene ID (Entrez) | 3421 |
Name | IDH3G, mitochondrial Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16071460
|
enQuireBio™
QP6194-EC-100UG |
100 μg |
530.00 €
100µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16091460
|
enQuireBio™
QP6194-EC-1MG |
1 mg |
2385.00 €
1mg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16081460
|
enQuireBio™
QP6194-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16001470
|
enQuireBio™
QP6194-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16011470
|
enQuireBio™
QP6194-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16021470
|
enQuireBio™
QP6194-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_004126.1 | |
IDH3G, mitochondrial Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
3421 | |
Research Use Only | |
IDH3G | |
Recombinant Protein | |
E. coli | |
FSEQTIPPSA KYGGRHTVTM IPGDGIGPEL MLHVKSVFRH ACVPVDFEEV HVSSNADEED IRNAIMAIRR NRVALKGNIE TNHNLPPSHK SRNNILRTSL DLYANVIHCK SLPGVVTRHK DIDILIVREN TEGEYSSLEH ESVAGVVESL KIITKAKSLR IAEYAFKLAQ ESGRKKVTAV HKANIMKLGD GLFLQCCREV AARYPQITFE NMIVDNTTMQ LVSRPQQFDV MVMPNLYGNI VNNVCAGLVG GPGLVAGANY GHVYAVFETA TRNTGKSIAN KNIANPTATL LASCMMLDHL KLHSYATSIR KAVLASMDNE NMHTPDIGGQ GTTSEAIQDV IRHIRVINGR AVEA | |
Greater than 90% as determined by SDS-PAGE. |