missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HSD11B2 Protein

A DNA sequence encoding the Homo sapiens (Human) HSD11B2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9185-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16097801

Quantity Price
1 1443.46 € / 500µg
3 1356.85 € / 500µg
Estimated Shipment
09-06-2021
Add to basket


Description

Description

Specifications

Specifications

NP_000187.3
HSD11B2 Protein
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
3291
Greater than 90% as determined by SDS-PAGE.
Research Use Only
HSD11B2
Recombinant Protein
Yeast
MERWPWPSGG AWLLVAARAL LQLLRSDLRL GRPLLAALAL LAALDWLCQR LLPPPAALAV LAAAGWIALS RLARPQRLPV ATRAVLITGC DSGFGKETAK KLDSMGFTVL ATVLELNSPG AIELRTCCSP RLRLLQMDLT KPGDISRVLE FTKAHTTSTG LWGLVNNAGH NEVVADAELS PVATFRSCME VNFFGALELT KGLLPLLRSS RGRIVTVGSP AGDMPYPCLG AYGTSKAAVA LLMDTFSCEL LPWGVKVSII QPGCFKTESV RNVGQWEKRK QLLLANLPQE LLQAYGKDYI EHLHGQFLHS LRLAMSDLTP VVDAITDALL AARPRRRYYP GQGLGLMYFI HYYLPEGLRR RFLQAFFISH CLPRALQPGQ PGTTPPQDAA QDPNLSPGPS PAVAR
Videos
SDS
Documents