missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Homeobox protein Hox-B4 Protein
A DNA sequence encoding the Homo sapiens (Human) Homeobox protein Hox-B4, was expressed in the hosts and tags indicated.
202.00 € - 2155.00 €
Specifications
Accession Number | NP_076920.1 |
---|---|
Gene ID (Entrez) | 3214 |
Name | Homeobox protein Hox-B4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16097791
|
enQuireBio™
QP9183-YE-100UG |
100 μg |
645.00 €
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16037801
|
enQuireBio™
QP9183-YE-500UG |
500 μg |
1740.00 €
500µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16007801
|
enQuireBio™
QP9183-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16027801
|
enQuireBio™
QP9183-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16047801
|
enQuireBio™
QP9183-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16017801
|
enQuireBio™
QP9183-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_076920.1 | |
Homeobox protein Hox-B4 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3214 | |
Research Use Only | |
HOXB4 | |
Recombinant Protein | |
Yeast | |
MAMSSFLINS NYVDPKFPPC EEYSQSDYLP SDHSPGYYAG GQRRESSFQP EAGFGRRAAC TVQRYAACRD PGPPPPPPPP PPPPPPPGLS PRAPAPPPAG ALLPEPGQRC EAVSSSPPPP PCAQNPLHPS PSHSACKEPV VYPWMRKVHV STVNPNYAGG EPKRSRTAYT RQQVLELEKE FHYNRYLTRR RRVEIAHALC LSERQIKIWF QNRRMKWKKD HKLPNTKIRS GGAAGSAGGP PGRPNGGPRA L | |
Greater than 90% as determined by SDS-PAGE. |