missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HLA-DQA2 Protein

A DNA sequence encoding the Homo sapiens (Human) HLA-DQA2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8816-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16046641

  • 2385.00 € / 1mg
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_064440.1
HLA-DQA2 Protein
Research Use Only
HLA-DQA2
Recombinant Protein
E. coli
EDIVADHVAS YGVNFYQSHG PSGQYTHEFD GDEEFYVDLE TKETVWQLPM FSKFISFDPQ SALRNMAVGK HTLEFMMRQS NSTAATNEVP EVTVFSKFPV TLGQPNTLIC LVDNIFPPVV NITWLSNGHS VTEGVSETSF LSKSDHSFFK ISYLTFLPSA DEIYDCKVEH WGLDEPLLKH WEPEIPAPMS E
Greater than 90% as determined by SDS-PAGE.
3118
1 mg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions