missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Heme-binding protein 1
A DNA sequence encoding the Homo sapiens (Human) Heme-binding protein 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8272-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_055135.1 | |
Heme-binding protein 1 | |
Research Use Only | |
HEBP2 | |
Recombinant Protein | |
E. coli | |
AEPLQPDPGA AEDAAAQAVE TPGWKAPEDA GPQPGSYEIR HYGPAKWVST SVESMDWDSA IQTGFTKLNS YIQGKNEKEM KIKMTAPVTS YVEPGSGPFS ESTITISLYI PSEQQFDPPR PLESDVFIED RAEMTVFVRS FDGFSSAQKN QEQLLTLASI LREDGKVFDE KVYYTAGYNS PVKLLNRNNE VWLIQKNEPT KENE | |
Greater than 90% as determined by SDS-PAGE. |
23593 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |