missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GRK2 / ADRBK1 Protein

A DNA sequence encoding the Homo sapiens (Human) GRK2 / ADRBK1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8340-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16094101

  • 493.03 € / 100µg

Estimated Shipment: 12-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_001610.2
GRK2 / ADRBK1 Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
156
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ADRBK1
Recombinant Protein
E. coli
ADLEAVLADV SYLMAMEKSK ATPAARASKK ILLPEPSIRS VMQKYLEDRG EVTFEKIFSQ KLGYLLFRDF CLNHLEEARP LVEFYEEIKK YEKLETEEER VARSREIFDS YIMKELLACS HPFSKSATEH VQGHLGKKQV PPDLFQPYIE EICQNLRGDV FQKFIESDKF TRFCQWKNVE LNIHLTMNDF SVHRIIGRGG FGEVYGCRKA DTGKMYAMKC
Videos
SDS
Documents
Certificates
Promotions

Promotions