missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Granzyme B / GZMB Protein

A DNA sequence encoding the Homo sapiens (Human) Granzyme B / GZMB, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6136-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16071080

  • 1740.00 € / 500µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_004122.2
Granzyme B / GZMB Protein
Research Use Only
GZMB
Recombinant Protein
Yeast
IIGGHEAKPH SRPYMAYLMI WDQKSLKRCG GFLIRDDFVL TAAHCWGSSI NVTLGAHNIK EQEPTQQFIP VKRPIPHPAY NPKNFSNDIM LLQLERKAKR TRAVQPLRLP SNKAQVKPGQ TCSVAGWGQT APLGKHSHTL QEVKMTVQED RKCESDLRHY YDSTIELCVG DPEIKKTSFK GDSGGPLVCN KVAQGIVSYG RNNGMPPRAC TKVSSFVHWI KKTMKRY
Greater than 90% as determined by SDS-PAGE.
3002
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions