missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GPD1 / Glycerolphosphate Dehydrogenase 1 Protein

A DNA sequence encoding the Homo sapiens (Human) GPD1 / Glycerolphosphate Dehydrogenase 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6111-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16090870

  • 530.00 € / 100µg
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001244128.1
GPD1 / Glycerolphosphate Dehydrogenase 1 Protein
Research Use Only
GPD1
Recombinant Protein
E. coli
MASKKVCIVG SGNWGSAIAK IVGGNAAQLA QFDPRVTMWV FEEDIGGKKL TEIINTQHEN VKYLPGHKLP PNVVAVPDVV QAAEDADILI FVVPHQFIGK ICDQLKGHLK ANATGISLIK GVDEGPNGLK LISEVIGERL GIPMSVLMGA NIASEVADEK FCETTIGCKD PAQGQLLKEL MQTPNFRITV VQEVDTVEIC GALKNVVAVG AGFCDGLGFG DNTKAAVIRL GLMEMIAFAK LFCSGPVSSA TFLESCGVAD LITTCYGGRN RKVAEAFART GKSIEQLEKE LLNGQKLQGP ETARELYSIL QHKGLVDKFP LFMAVYKVCY EGQPVGEFIH CLQNHPEHM
Greater than 90% as determined by SDS-PAGE.
2819
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions