missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glutathione S-transferase A4 Protein
A DNA sequence encoding the Homo sapiens (Human) Glutathione S-transferase A4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6124-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001503.1 | |
Glutathione S-transferase A4 Protein | |
Research Use Only | |
GSTA4 | |
Recombinant Protein | |
E. coli | |
MAARPKLHYP NGRGRMESVR WVLAAAGVEF DEEFLETKEQ LYKLQDGNHL LFQQVPMVEI DGMKLVQTRS ILHYIADKHN LFGKNLKERT LIDMYVEGTL DLLELLIMHP FLKPDDQQKE VVNMAQKAII RYFPVFEKIL RGHGQSFLVG NQLSLADVIL LQTILALEEK IPNILSAFPF LQEYTVKLSN IPTIKRFLEP GSKKKPPPDE IYVRTVYNIF RP | |
Greater than 90% as determined by SDS-PAGE. |
2941 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |