missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ESAM / Endothelial Cell Adhesion Molecule Protein

A DNA sequence encoding the Homo sapiens (Human) ESAM / Endothelial Cell Adhesion Molecule, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7960-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16071841

Quantity Price
1 493.03 € / 100µg
6 463.44 € / 100µg
Estimated Shipment: 17-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_620411.2
ESAM / Endothelial Cell Adhesion Molecule Protein
100 μg
Not determined.
Extracellular Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
90952
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ESAM
Recombinant Protein
E. coli
QLQLHLPANR LQAVEGGEVV LPAWYTLHGE VSSSQPWEVP FVMWFFKQKE KEDQVLSYIN GVTTSKPGVS LVYSMPSRNL SLRLEGLQEK DSGPYSCSVN VQDKQGKSRG HSIKTLELNV LVPPAPPSCR LQGVPHVGAN VTLSCQSPRS KPAVQYQWDR QLPSFQTFFA PALDVIRGSL SLTNLSSSMA GVYVCKAHNE VGTAQCNVTL EVSTGPGAA
Videos
SDS
Documents
Certificates
Promotions

Promotions