missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase TRIM9 Protein

A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase TRIM9, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8167-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16093011

  • 2248.90 € / 1mg

Estimated Shipment: 10-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_055978.4
E3 ubiquitin-protein ligase TRIM9 Protein
1 mg
Not determined.
Full Length of isoform 5
Human
His-SUMO
Tris-based buffer, 50% glycerol
114088
Greater than 90% as determined by SDS-PAGE.
Research Use Only
TRIM9
Recombinant Protein
E. coli
MEEMEEELKC PVCGSFYREP IILPCSHNLC QACARNILVQ TPESESPQSH RAAGSGVSDY DYLDLDKMSL YSEADSGYGS YGGFASAPTT PCQKSPNGVR VFPPAMPPPA THLSPALAPV PRNSCITCPQ CHRSLILDDR GLRGFPKNRV LEGVIDRYQQ SKAAALKCQL CEKAPKEATV MCEQCDVFYC DPCRLRCHPP RGPLAKHRLV PPAQGRVSRR LSPRKVSTCT DHELENHSMY CVQCKMPVCY QCLEEGKHSS HEVKALGAMW KLHKSQLSQA LNGLSDRAKE AKEFLVQLRN MVQQIQENSV EFEACLVAQC DALIDALNRR KAQLLARVNK EHEHKLKVVR DQISHCTVKL RQTTGLMEYC LEVIKENDPS GFLQISDALI RRVHLTEDQW GKGTLTPRMT TDFDLSLDNS PLLQSIHQLD FVQVKASSPV PATPILQLEE CCTHNNSATL SWKQPPLSTV PADGYILELD DGNGGQFREV YVGKETMCTV DGLHFNSTYN ARVKAFNKTG VSPYSKTLVL QTSEGKALQQ YPSERELRGI
Videos
SDS
Documents
Certificates
Promotions

Promotions