missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Dynactin subunit 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Dynactin subunit 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7570-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16089460

  • 2385.00 € / 1mg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001128512.1
Dynactin subunit 1 Protein
Research Use Only
DCTN1
Recombinant Protein
E. coli
PSKEEEGLRA QVRDLEEKLE TLRLKRAEDK AKLKELEKHK IQLEQVQEWK SKMQEQQADL QRRLKEARKE AKEALEAKER YMEEMADTAD AIEMATLDKE MAEERAESLQ QEVEALKERV DELTTDLEIL KAEIEEKGSD GAASSYQLKQ LEEQNARLKD ALVRMRDLSS SEKQEHVKLQ KLMEKKNQEL EVVRQQRERL QEELSQAEST IDELKEQVDA ALGAEEMVEM LTDRNLNLEE KVRELRETVG DLEAMNEMND ELQENARETE LELREQLDMA GARVREAQKR VEAAQETVAD YQQTIKKYRQ LTAHLQDVNR ELTNQQEASV ERQQQ
Greater than 84.6% as determined by SDS-PAGE.
1639
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions