missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Dynactin subunit 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Dynactin subunit 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7570-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001128512.1 | |
Dynactin subunit 1 Protein | |
Research Use Only | |
DCTN1 | |
Recombinant Protein | |
E. coli | |
PSKEEEGLRA QVRDLEEKLE TLRLKRAEDK AKLKELEKHK IQLEQVQEWK SKMQEQQADL QRRLKEARKE AKEALEAKER YMEEMADTAD AIEMATLDKE MAEERAESLQ QEVEALKERV DELTTDLEIL KAEIEEKGSD GAASSYQLKQ LEEQNARLKD ALVRMRDLSS SEKQEHVKLQ KLMEKKNQEL EVVRQQRERL QEELSQAEST IDELKEQVDA ALGAEEMVEM LTDRNLNLEE KVRELRETVG DLEAMNEMND ELQENARETE LELREQLDMA GARVREAQKR VEAAQETVAD YQQTIKKYRQ LTAHLQDVNR ELTNQQEASV ERQQQ | |
Greater than 84.6% as determined by SDS-PAGE. |
1639 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |