missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Dynactin subunit 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Dynactin subunit 1, was expressed in the hosts and tags indicated.
160.00 € - 2385.00 €
Specifications
Accession Number | NP_001128512.1 |
---|---|
Gene ID (Entrez) | 1639 |
Name | Dynactin subunit 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16069460
|
enQuireBio™
QP7570-EC-100UG |
100 μg |
530.00 €
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16089460
|
enQuireBio™
QP7570-EC-1MG |
1 mg |
2385.00 €
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16079460
|
enQuireBio™
QP7570-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16099460
|
enQuireBio™
QP7570-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16009470
|
enQuireBio™
QP7570-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16019470
|
enQuireBio™
QP7570-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001128512.1 | |
Dynactin subunit 1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
1639 | |
Research Use Only | |
DCTN1 | |
Recombinant Protein | |
E. coli | |
PSKEEEGLRA QVRDLEEKLE TLRLKRAEDK AKLKELEKHK IQLEQVQEWK SKMQEQQADL QRRLKEARKE AKEALEAKER YMEEMADTAD AIEMATLDKE MAEERAESLQ QEVEALKERV DELTTDLEIL KAEIEEKGSD GAASSYQLKQ LEEQNARLKD ALVRMRDLSS SEKQEHVKLQ KLMEKKNQEL EVVRQQRERL QEELSQAEST IDELKEQVDA ALGAEEMVEM LTDRNLNLEE KVRELRETVG DLEAMNEMND ELQENARETE LELREQLDMA GARVREAQKR VEAAQETVAD YQQTIKKYRQ LTAHLQDVNR ELTNQQEASV ERQQQ | |
Greater than 84.6% as determined by SDS-PAGE. |