missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DNA repair protein XRCC4 Protein

A DNA sequence encoding the Homo sapiens (Human) DNA repair protein XRCC4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7540-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16089260

  • 725.49 € / 100µg

Estimated Shipment: 10-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_001304941.1
DNA repair protein XRCC4 Protein
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
7518
Greater than 80% as determined by SDS-PAGE.
Research Use Only
XRCC4
Recombinant Protein
E. coli
MERKISRIHL VSEPSITHFL QVSWEKTLES GFVITLTDGH SAWTGTVSES EISQEADDMA MEKGKYVGEL RKALLSGAGP ADVYTFNFSK ESCYFFFEKN LKDVSFRLGS FNLEKVENPA EVIRELICYC LDTIAENQAK NEHLQKENER LLRDWNDVQG RFEKCVSAKE ALETDLYKRF ILVLNEKKTK IRSLHNKLLN AAQEREKDIK QEGETAICSE MTADRDPVYD ESTDEESENQ TDLSGLASAA VSKDDSIISS LDVTDIAPSR KRRQRMQRNL GTEPKMAPQE NQLQEKENSR PDSSLPETSK KEHISAENMS LETLRNSSPE DLFDEI
Videos
SDS
Documents
Certificates
Promotions

Promotions