missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNA ligase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) DNA ligase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6308-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_000225.1 | |
DNA ligase 1 Protein | |
Research Use Only | |
LIG1 | |
Recombinant Protein | |
E. coli | |
MQRSIMSFFH PKKEGKAKKP EKEASNSSRE TEPPPKAALK EWNGVVSESD SPVKRPGRKA ARVLGSEGEE EDEALSPAKG QKPALDCSQV SPPRPATSPE NNASLSDTSP MDSSPSGIPK RRTARKQLPK RTIQEVLEEQ SEDEDREAKR KKEEEEEETP KESLTEAEVA TEKEGEDGDQ PTTPPKPLKT SKAETPTESV SEPEVATKQE LQEEEEQTKP PRRAPKTLSS FFTPRKPAVK KEVKEEEPGA PGKEGAAEGP LDPSGYNPAK NNYHPVEDAC WKPGQKVPYL AVARTFEKIE EVSARLRMVE TLSNLLRSVV ALSPPDLLPV LYLSLNHLGP PQQGLELGVG DGVLLKAVAQ ATGRQLESVR AEAAEKGDVG LVAENSRSTQ RLMLPPPPLT ASGVFSKFRD IARLTGSAST AKKIDIIKGL FVACRHSEAR FIARSLSGRL RLGLAEQSVL AALSQAVSLT PPGQEFPPAM VDAGKGKTAE ARKTWLEEQG MILKQTFCEV PDLDRIIPVL LEHGLERLPE HCKLSPGIPL KPMLAHPTRG ISEVLKRFEE AAFTCEYKYD GQRAQIHALE GGEVKIFSRN QEDNTGKYPD IISRIPKIKL PSVTSFILDT EAVAWDREKK QIQPFQVLTT RKRKEVDASE IQVQVCLYAF DLIYLNGESL VREPLSRRRQ LLRENFVETE GEFVFATSLD TKDIEQIAEF LEQSVKDSCE GLMVKTLDVD ATYEIAKRSH NWLKLKKDYL DGVGDTLDLV VIGAYLGRGK RAGRYGGFLL ASYDEDSEEL QAICKLGTGF SDEELEEHHQ SLKALVLPSP RPYVRIDGAV IPDHWLDPSA VWEVKCADLS LSPIYPAARG LVDSDKGISL RFPRFIRVRE DKQPEQATTS AQVACLYRKQ SQIQNQQGED | |
Greater than 90% as determined by SDS-PAGE. |
3978 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |