missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human d2hgdh, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) d2hgdh, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7919-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16091581

Quantity Price
1 493.03 € / 100µg
6 463.44 € / 100µg
Estimated Shipment: 17-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_001274178.1
d2hgdh, mitochondrial Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
728294
Greater than 90% as determined by SDS-PAGE.
Research Use Only
d2hgdh
Recombinant Protein
E. coli
LRGAPGAAGS WGRPVGPLAR RGCCSAPGTP EVPLTRERYP VRRLPFSTVS KQDLAAFERI VPGGVVTDPE ALQAPNVDWL RTLRGCSKVL LRPRTSEEVS HILRHCHERN LAVNPQGGNT GMVGGSVPVF DEIILSTARM NRVLSFHSVS GILVCQAGCV LEELSRYVEE RDFIMPLDLG AKGSCHIGGN VATNAGGLRF LRYGSLHGTV LGLEVVLADG TVLDCLTSLR KDNTGYDLKQ LFIGSEGTLG IITTVSILCP PKPRAVNVAF LGCPGFAEVL QTFSTCKGML GEILSAFEFM DAVCMQLVGR HLHLASPVQE SPFYVLIETS GSNAGHDAEK LGHFLEHALG SGLVTDGTMA TDQRKVKMLW ALRERITEAL SRDGYVYKYD LSLPVERLYD IVTDLRARLG PHAKHVVGYG HLGDGNLHLN VTAEAFSPSL LAALEPHVYE WTAGQQGSVS AEHGVGFRKR DVLGYSKPPG ALQLMQQLKA LLDPKGILNP YKTLPSQA
Videos
SDS
Documents
Certificates
Promotions

Promotions