missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Corticotropin-releasing factor receptor 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Corticotropin-releasing factor receptor 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9080-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16097581

Quantity Price
1 2300.65 € / 1mg
3 2163.76 € / 1mg
Estimated Shipment
09-06-2021
Add to basket


Description

Description

Specifications

Specifications

NP_001138618.1
Corticotropin-releasing factor receptor 1 Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
104909134
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CRHR1
Recombinant Protein
Yeast
ASLQDQHCES LSLASNISGL QCNASVDLIG TCWPRSPAGQ LVVRPCPAFF YGVRYNTTNN GYRECLANGS WAARVNYSEC QEILNEEKKS KVHYHVAVI
Videos
SDS
Documents