missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Collagen alpha-6(VI) chain Protein
A DNA sequence encoding the Homo sapiens (Human) Collagen alpha-6(VI) chain, was expressed in the hosts and tags indicated.
160.00 € - 2385.00 €
Specifications
Accession Number | NP_001096078.1 |
---|---|
Gene ID (Entrez) | 131873 |
Name | Collagen alpha-6(VI) chain Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16068830
|
enQuireBio™
QP7431-EC-100UG |
100 μg |
530.00 €
100µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16088830
|
enQuireBio™
QP7431-EC-1MG |
1 mg |
2385.00 €
1mg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16078830
|
enQuireBio™
QP7431-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098830
|
enQuireBio™
QP7431-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16008840
|
enQuireBio™
QP7431-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16018840
|
enQuireBio™
QP7431-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001096078.1 | |
Collagen alpha-6(VI) chain Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
131873 | |
Research Use Only | |
COL6A6 | |
Recombinant Protein | |
E. coli | |
VDTEEADIYL LIDGSGSTQA TDFHEMKTFL SEVVGMFNIA PHKVRVGAVQ YADSWDLEFE INKYSNKQDL GKAIENIRQM GGNTNTGAAL NFTLSLLQKA KKQRGNKVPC HLVVLTNGMS KDSILEPANR LREEHIRVYA IGIKEANQTQ LREIAGEEKR VYYVHDFDAL KDIRNQVVQE ICTEEACKEM KADIMFLVDS SGSIGPENFS KMKTFMKNLV SKSQIGPDRV QIGVVQFSDI NKEEFQLNRF MSQSDISNAI DQMAHIGQTT LTGSALSFVS QYFSPTKGAR PNIRKFLILI TDGEAQDIVK EPAVVLRQEG VIIYSVGVFG SNVTQLEEIS GRPEMVFYVE NFDILQRIED DLVFGICSPR EECKRIEVLD VVFVIDSSGS IDYDEYNIMK DFMIGLVKKA DVGKNQVRFG ALKYADDPEV LFYLDDFGTK LEVISVLQND QAMGGSTYTA EALGFSDHMF TEARGSRLNK GVPQVLIVIT DGESHDADKL NATAKALRDK GILVLAVGID GANPVELLAM AGSSDKYFFV ETFGGLKGIF SDV | |
Greater than 90% as determined by SDS-PAGE. |