missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Checkpoint protein HUS1 Protein

A DNA sequence encoding the Homo sapiens (Human) Checkpoint protein HUS1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6186-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16091410

  • 493.03 € / 100µg

Estimated Shipment: 10-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_004498.1
Checkpoint protein HUS1 Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
3364
Greater than 90% as determined by SDS-PAGE.
Research Use Only
HUS1
Recombinant Protein
E. coli
KFRAKIVDGA CLNHFTRISN MIAKLAKTCT LRISPDKLNF ILCDKLANGG VSMWCELEQE NFFNEFQMEG VSAENNEIYL ELTSENLSRA LKTAQNARAL KIKLTNKHFP CLTVSVELLS MSSSSRIVTH DIPIKVIPRK LWKDLQEPVV PDPDVSIYLP VLKTMKSVVE KMKNISNHLV IEANLDGELN LKIETELVCV TTHFKDLGNP PLASESTHED RNVEHMAEVH IDIRKLLQFL AGQQVNPTKA LCNIVNNKMV HFDLLHEDVS LQYFIPALS
Videos
SDS
Documents
Certificates
Promotions

Promotions