missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cell death activator CIDE-3 Protein

A DNA sequence encoding the Homo sapiens (Human) Cell death activator CIDE-3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7912-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16091541

  • 2248.90 € / 1mg

Estimated Shipment: 10-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_001186480.1
Cell death activator CIDE-3 Protein
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
63924
Greater than 80% as determined by SDS-PAGE.
Research Use Only
CIDEC
Recombinant Protein
E. coli
MEYAMKSLSL LYPKSLSRHV SVRTSVVTQQ LLSEPSPKAP RARPCRVSTA DRSVRKGIMA YSLEDLLLKV RDTLMLADKP FFLVLEEDGT TVETEEYFQA LAGDTVFMVL QKGQKWQPPS EQGTRHPLSL SHKPAKKIDV ARVTFDLYKL NPQDFIGCLN VKATFYDTYS LSYDLHCCGA KRIMKEAFRW ALFSMQATGH VLLGTSCYLQ QLLDATEEGQ PPKGKASSLI PTCLKILQ
Videos
SDS
Documents
Certificates
Promotions

Promotions