missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD52 / CDW52 Protein

A DNA sequence encoding the Homo sapiens (Human) CD52 / CDW52, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9049-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16097531

  • 920.00 € / 100µg
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001794.2
CD52 / CDW52 Protein
Research Use Only
CD52
Recombinant Protein
Yeast
GQNDTSQTSS PS
Greater than 90% as determined by SDS-PAGE.
1043
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions