missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD3e / CD3 epsilon Protein
A DNA sequence encoding the Homo sapiens (Human) CD3e / CD3 epsilon, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9044-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000724.1 | |
CD3e / CD3 epsilon Protein | |
Research Use Only | |
CD3E | |
Recombinant Protein | |
Yeast | |
DGNEEMGGIT QTPYKVSISG TTVILTCPQY PGSEILWQHN DKNIGGDEDD KNIGSDEDHL SLKEFSELEQ SGYYVCYPRG SKPEDANFYL YLRARVCENC MEMD | |
Greater than 90% as determined by SDS-PAGE. |
916 | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |