missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD300LB / LMIR5 / CMRF35-A2 Protein
A DNA sequence encoding the Homo sapiens (Human) CD300LB / LMIR5 / CMRF35-A2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9043-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_777552.3 | |
CD300LB / LMIR5 / CMRF35-A2 Protein | |
Research Use Only | |
CD300LB | |
Recombinant Protein | |
Yeast | |
IQGPESVRAP EQGSLTVQCH YKQGWETYIK WWCRGVRWDT CKILIETRGS EQGEKSDRVS IKDNQKDRTF TVTMEGLRRD DADVYWCGIE RRGPDLGTQV KVIVDPEGAA STTASSPTNS NMAVFIGSHK RNHY | |
Greater than 90% as determined by SDS-PAGE. |
124599 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |