missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Casein kinase I isoform alpha-like Protein
A DNA sequence encoding the Homo sapiens (Human) Casein kinase I isoform alpha-like, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7904-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_660204.2 | |
Casein kinase I isoform alpha-like Protein | |
Research Use Only | |
CSNK1A1L | |
Recombinant Protein | |
E. coli | |
MTNNSGSKAE LVVGGKYKLV RKIGSGSFGD VYLGITTTNG EEVAVKLESQ KVKHPQLLYE SKLYTILQGG VGIPHMHWYG QEKDNNVLVM DLLGPSLEDL FNFCSRRFTM KTVLMLADQM ISRIEYVHTK NFLHRDIKPD NFLMGTGRHC NKLFLIDFGL AKKYRDNRTR QHIPYREDKH LIGTVRYASI NAHLGIEQSR RDDMESLGYV FMYFNRTSLP WQGLKAMTKK QKYEKISEKK MSTPVEVLCK GFPAEFAMYL NYCRGLRFEE VPDYMYLRQL FRILFRTLNH QYDYTFDWTM LKQKAAQQAA SSSGQGQQAQ TQTGKQTEKN KNNVKDN | |
Greater than 80% as determined by SDS-PAGE. |
122011 | |
1 mg | |
Not determined. | |
Full Length of BC028723 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |