missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CALML5 / CLSP Protein

A DNA sequence encoding the Homo sapiens (Human) CALML5 / CLSP, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8302-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16093881

  • 2385.00 € / 1mg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_059118.2
CALML5 / CLSP Protein
Research Use Only
CALML5
Recombinant Protein
E. coli
AGELTPEEEA QYKKAFSAVD TDGNGTINAQ ELGAALKATG KNLSEAQLRK LISEVDSDGD GEISFQEFLT AAKKARAGLE DLQVAFRAFD QDGDGHITVD ELRRAMAGLG QPLPQEELDA MIREADVDQD GRVNYEEFAR MLAQE
Greater than 90% as determined by SDS-PAGE.
51806
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions