missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Calcium-dependent phospholipase A2 Protein
A DNA sequence encoding the Homo sapiens (Human) Calcium-dependent phospholipase A2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9333-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000920.1 | |
Calcium-dependent phospholipase A2 Protein | |
Research Use Only | |
PLA2G5 | |
Recombinant Protein | |
Yeast | |
GLLDLKSMIE KVTGKNALTN YGFYGCYCGW GGRGTPKDGT DWCCWAHDHC YGRLEEKGCN IRTQSYKYRF AWGVVTCEPG PFCHVNLCAC DRKLVYCLKR NLRSYNPQYQ YFPNILCS | |
Greater than 90% as determined by SDS-PAGE. |
5322 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |