missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BANK1 Protein

A DNA sequence encoding the Homo sapiens (Human) BANK1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9855-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16078851

  • 2765.00 € / 1mg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001077376.2
BANK1 Protein
Research Use Only
BANK1
Recombinant Protein
Yeast
MLPAAPGKGL GSPDPAPCGP APPGNTKDII MIYEEDAEEW ALYLTEVFLH VVKREAILLY RLENFSFRHL ELLNLTSYKC KLLILSNSLL RDLTPKKCQF LEKILHSPKS VVTLLCGVKS SDQLYELLNI SQSRWEISTE QEPEDYISVI QSIIFKDSED YFEVNIPTDL RAKHSGEISE RKEIEELSEA SRNTIPLAVV LPTEIPCENP GEIFIILRDE VIGDTVEVEF TSSNKRIRTR PALWNKKVWC MKALEFPAGS VHVNVYCDGI VKATTKIKYY PTAKAKECLF RMADSGESLC QNSIEELDGV LTSIFKHEIP YYEFQSLQTE ICSQNKYTHF KELPTLLHCA AKFGLKNLAI HLLQCSGATW ASKMKNMEGS DPAHIAERHG HKELKKIFED FSIQEIDINN EQENDYEEDI ASFSTYIPST QNPAFHHESR KTYGQSADGA EANEMEGEGK QNGSGMETKH SPLEVGSESS EDQYDDLYVF IPGADPENNS QEPLMSSRPP LPPPRPVANA FQLERPHFTL PGTMVEGQME RSQNWGHPGV RQETGDEPKG EKEKKEEEKE QEEEEDPYTF AEIDDSEYDM ILANLSIKKK TGSRSFIINR PPAPTPRPTS IPPKEETTPY IAQVFQQKTA RRQSDDDKFC GLPKKQDRAR IESPAFSTLR GCLTDGQEEL ILLQEKVKNG KMSMDEALEK FKHWQMGKSG LEMIQQEKLR QLRDCIIGKR PEEENVYNKL TIVHHPGGKE TAHNENKFYN VHFSNKLPAR PQVEKEFGFC CKKDH
Greater than 90% as determined by SDS-PAGE.
55024
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions