missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Angiopoietin-like protein 1
A DNA sequence encoding the Homo sapiens (Human) Angiopoietin-like protein 8, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7768-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_061157.3 | |
Angiopoietin-like protein 1 | |
Research Use Only | |
C19orf80 | |
Recombinant Protein | |
E. coli | |
APMGGPELAQ HEELTLLFHG TLQLGQALNG VYRTTEGRLT KARNSLGLYG RTIELLGQEV SRGRDAAQEL RASLLETQME EDILQLQAEA TAEVLGEVAQ AQKVLRDSVQ RLEVQLRSAW LGPAYREFEV LKAHADKQSH ILWALTGHVQ RQRREMVAQQ HRLRQIQERL HTAALPA | |
Greater than 90% as determined by SDS-PAGE. |
55908 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |