missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human AGO2 / Argonaute 2 / EIF2C2 Protein

A DNA sequence encoding the Homo sapiens (Human) AGO2 / Argonaute 2 / EIF2C2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8243-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16083511

Quantity Price
1 595.74 € / 100µg
5 560.65 € / 100µg
Estimated Shipment: 18-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_001158095.1
AGO2 / Argonaute 2 / EIF2C2 Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
27161
Greater than 80% as determined by SDS-PAGE.
Research Use Only
AGO2
Recombinant Protein
Yeast
LVVVILPGKT PVYAEVKRVG DTVLGMATQC VQMKNVQRTT PQTLSNLCLK INVKLGGVNN ILLPQGRPPV FQQPVIFLGA DVTHPPAGDG KKPSIAAVVG SMDAHPNRYC ATVRVQQHRQ EIIQDLAAMV RELLIQFYKS TRFKPTRIIF YRDGVSEGQF QQVLHHELLA IREACIKLEK DYQPGITFIV VQKRHHTRLF CTDKNERVGK SGNIPAGTTV DTKITHPTEF DFYLCSHAGI QGTSRPSHYH VLWDDNRFSS DELQILTYQL CHTYVRCTRS VSIPAPAYYA HLVAFRARYH LV
Videos
SDS
Documents
Certificates
Promotions

Promotions