missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ADAM15 / MDC15 Protein
A DNA sequence encoding the Homo sapiens (Human) ADAM15 / MDC15, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8390-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001248393.1 | |
ADAM15 / MDC15 Protein | |
Research Use Only | |
ADAM15 | |
Recombinant Protein | |
E. coli | |
DVVTETKTVE LVIVADHSEA QKYRDFQHLL NRTLEVALLL DTFFRPLNVR VALVGLEAWT QRDLVEISPN PAVTLENFLH WRRAHLLPRL PHDSAQLVTG TSFSGPTVGM AIQNSICSPD FSGGVNMDHS TSILGVASSI AHELGHSLGL DHDLPGNSCP CPGPAPAKTC IMEASTDFLP GLNFSNCSRR ALEKALLDGM GSCLFERLPS LPPMAAFCGN MFVEPGEQCD CGFLDDCVDP CCDSLT | |
Greater than 90% as determined by SDS-PAGE. |
8751 | |
1 mg | |
Not determined. | |
Extracellular Domain | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |