missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human 60S ribosomal protein L14 Protein

A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L14, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8323-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16054011

  • 2365.00 € / 1mg

Estimated Shipment: 18-11-2022
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001030168.1
60S ribosomal protein L14 Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
9045
Greater than 90% as determined by SDS-PAGE.
Research Use Only
RPL14
Recombinant Protein
E. coli
VFRRFVEVGR VAYVSFGPHA GKLVAIVDVI DQNRALVDGP CTQVRRQAMP FKCMQLTDFI LKFPHSAHQK YVRQAWQKAD INTKWAATRW AKKIEARERK AKMTDFDRFK VMKAKKMRNR IIKNEVKKLQ KAALLKASPK KAPGTKGTAA AAAAAAAAKV PAKKITAASK KAPAQKVPAQ KATGQKAAPA PKAQKGQKAP AQKAPAPKAS GKK
Videos
SDS
Documents
Certificates
Promotions

Promotions