missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant H. pylori Vacuolating cytotoxin autotransporter Protein
A DNA sequence encoding the Helicobacter pylori (strain ATCC 700392 / 26695) (Campylobacter pylori) Vacuolating cytotoxin autotransporter, was expressed in the hosts and tags indicated. option, abo
309.00 € - 3405.00 €
Specifications
Accession Number | NP_207680.1 |
---|---|
Gene ID (Entrez) | 899415 |
Name | Vacuolating cytotoxin autotransporter Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16057790
|
enQuireBio™
QP7226-EC-100UG |
100 μg |
985.00 €
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16077790
|
enQuireBio™
QP7226-EC-1MG |
1 mg |
3405.00 €
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16067790
|
enQuireBio™
QP7226-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087790
|
enQuireBio™
QP7226-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16097790
|
enQuireBio™
QP7226-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16007800
|
enQuireBio™
QP7226-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_207680.1 | |
Vacuolating cytotoxin autotransporter Protein | |
Not determined. | |
Full Length | |
H. pylori | |
His-B2M | |
Tris-based buffer, 50% glycerol |
899415 | |
Research Use Only | |
vacA | |
Recombinant Protein | |
E. coli | |
KYEKPTNVWA NAIGGTSLNN GGNASLYGTS AGVDAYLNGE VEAIVGGFGS YGYSSFNNQA NSLNSGANNT NFGVYSRIFA NQHEFDFEAQ GALGSDQSSL NFKSALLRDL NQSYNYLAYS AATRASYGYD FAFFRNALVL KPSVGVSYNH LGSTNFKSNS NQVALKNGSS SQHLFNASAN VEARYYYGDT SYFYMNAGVL QEFANFGSSN AVSLNTFKVN AAHNPLSTHA RVMMGGELKL AKEVFLNLGF VYLHNLISNI GHFASNLGMR YSF | |
Greater than 90% as determined by SDS-PAGE. |