missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Trigger factor Protein

A DNA sequence encoding the Escherichia coli (strain K12) Trigger factor, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9773-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16068691

  • 1037.95 € / 100µg

Estimated Shipment: 10-01-2022
Add to basket

Description

Description

Specifications

Specifications

WP_001198385.1
Greater than 90% as determined by SDS-PAGE.
Research Use Only
tig
Recombinant Protein
Yeast
MQVSVETTQG LGRRVTITIA ADSIETAVKS ELVNVAKKVR IDGFRKGKVP MNIVAQRYGA SVRQDVLGDL MSRNFIDAII KEKINPAGAP TYVPGEYKLG EDFTYSVEFE VYPEVELQGL EAIEVEKPIV EVTDADVDGM LDTLRKQQAT WKEKDGAVEA EDRVTIDFTG SVDGEEFEGG KASDFVLAMG QGRMIPGFED GIKGHKAGEE FTIDVTFPEE YHAENLKGKA AKFAINLKKV EERELPELTA EFIKRFGVED GSVEGLRAEV RKNMERELKS AIRNRVKSQA IEGLVKANDI DVPAALIDSE IDVLRRQAAQ RFGGNEKQAL ELPRELFEEQ AKRRVVVGLL LGEVIRTNEL KADEERVKGL IEEMASAYED PKEVIEFYSK NKELMDNMRN VALEEQAVEA VLAKAKVTEK ETTFNELMN
Trigger factor Protein
100 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
Videos
SDS
Documents
Certificates
Promotions

Promotions