missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Thioredoxin-2 Protein

A DNA sequence encoding the Escherichia coli (strain K12) Thioredoxin-2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7012-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16087030

  • 1037.95 € / 100µg

Estimated Shipment: 10-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_417077.1
Thioredoxin-2 Protein
100 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
947062
Greater than 90% as determined by SDS-PAGE.
Research Use Only
trxC
Recombinant Protein
Yeast
MNTVCTHCQA INRIPDDRIE DAAKCGRCGH DLFDGEVINA TGETLDKLLK DDLPVVIDFW APWCGPCRNF APIFEDVAQE RSGKVRFVKV NTEAERELSS RFGIRSIPTI MIFKNGQVVD MLNGAVPKAP FDSWLNESL
Videos
SDS
Documents
Certificates
Promotions

Promotions