missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Ribose-5-phosphate isomerase A Protein

A DNA sequence encoding the Escherichia coli (strain K12) Ribose-5-phosphate isomerase A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7507-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16069070

  • 1365.00 € / 100µg
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_417389.1
Ribose-5-phosphate isomerase A Protein
Research Use Only
rpiA
Recombinant Protein
Yeast
MTQDELKKAV GWAALQYVQP GTIVGVGTGS TAAHFIDALG TMKGQIEGAV SSSDASTEKL KSLGIHVFDL NEVDSLGIYV DGADEINGHM QMIKGGGAAL TREKIIASVA EKFICIADAS KQVDILGKFP LPVEVIPMAR SAVARQLVKL GGRPEYRQGV VTDNGNVILD VHGMEILDPI AMENAINAIP GVVTVGLFAN RGADVALIGT PDGVKTIVK
Greater than 90% as determined by SDS-PAGE.
947407
100 μg
Not determined.
Full Length
E. coli
Untagged
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions