missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Pyruvate dehydrogenase [ubiquinone] Protein
A DNA sequence encoding the Escherichia coli (strain K12) Pyruvate dehydrogenase [ubiquinone], was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8913-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_415392.1 | |
Pyruvate dehydrogenase [ubiquinone] Protein | |
Research Use Only | |
poxB | |
Recombinant Protein | |
E. coli | |
MKQTVAAYIA KTLESAGVKR IWGVTGDSLN GLSDSLNRMG TIEWMSTRHE EVAAFAAGAE AQLSGELAVC AGSCGPGNLH LINGLFDCHR NHVPVLAIAA HIPSSEIGSG YFQETHPQEL FRECSHYCEL VSSPEQIPQV LAIAMRKAVL NRGVSVVVLP GDVALKPAPE GATMHWYHAP QPVVTPEEEE LRKLAQLLRY SSNIALMCGS GCAGAHKELV EFAGKIKAPI VHALRGKEHV EYDNPYDVGM TGLIGFSSGF HTMMNADTLV LLGTQFPYRA FYPTDAKIIQ IDINPASIGA HSKVDMALVG DIKSTLRALL PLVEEKADRK FLDKALEDYR DARKGLDDLA KPSEKAIHPQ YLAQQISHFA ADDAIFTCDV GTPTVWAARY LKMNGKRRLL GSFNHGSMAN AMPQALGAQA TEPERQVVAM CGDGGFSMLM GDFLSVVQMK LPVKIVVFNN SVLGFVAMEM KAGGYLTDGT ELHDTNFARI AEACGITGIR VEKASEVDEA LQRAFSIDGP VLVDVVVAKE ELAIPPQIKL EQAKGFSLYM LRAIISGRGD EVIELAKTNW LR | |
Greater than 90% as determined by SDS-PAGE. |
946132 | |
100 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |