missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Phosphoribosylamine--glycine ligase Protein

A DNA sequence encoding the Escherichia coli O157:H7 Phosphoribosylamine--glycine ligase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7954-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16091811

  • 3405.00 € / 1mg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_312955.1
Phosphoribosylamine--glycine ligase Protein
Research Use Only
purD
Recombinant Protein
E. coli
MKVLVIGNGG REHALAWKAA QSPLVETVFV APGNAGTALE PTLQNVAIGV TDIPALLDFA QNEKVDLTIV GPEAPLVKGV VDTFRAAGMK IFGPTAGAAQ LEGSKAFTKD FLARHNIPTA EYQNFTEVEP ALAYLREKGA PIVIKADGLA AGKGVIVAMT LEEAEAAVHD MLAGNAFGDA GHRIVIEEFL DGEEASFIVM VDGEHVLPMA TSQDHKRVGD KDTGPNTGGM GAYSPAPVVT DDVHQRTMER IIWPTVKGMA SEGNTYTGFL YAGLMIDKQG NPKVIEFNCR FGDPETQPIM LRMKSDLVEL CLAACEGKLD EKTSEWDERA SLGVVMAAGG YPGDYRTGDV IHGLPLEEVE DGKVFHAGTK LADDEQVVTS GGRVLCVTAL GHTVAEAQKR AYALMTDIHW DDCFCRKDIG WRAIEREQN
Greater than 90% as determined by SDS-PAGE.
914922
1 mg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions