missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Ornithine decarboxylase, inducible Protein

A DNA sequence encoding the Escherichia coli (strain K12) Ornithine decarboxylase, inducible, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8883-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16096961

Quantity Price
1 810.66 € / 100µg
5 762.51 € / 100µg
Estimated Shipment
09-06-2021
Add to basket


Description

Description

Specifications

Specifications

NP_415220.1
Ornithine decarboxylase, inducible Protein
100 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
945297
Greater than 90% as determined by SDS-PAGE.
Research Use Only
speF
Recombinant Protein
E. coli
MSKLKIAVSD SCPDCFTTQR ECIYINESRN IDVAAIVLSL NDVTCGKLDE IDATGYGIPV FIATENQERV PAEYLPRISG VFENCESRRE FYGRQLETAA SHYETQLRPP FFRALVDYVN QGNSAFDCPG HQGGEFFRRH PAGNQFVEYF GEALFRADLC NADVAMGDLL IHEGAPCIAQ QHAAKVFNAD KTYFVLNGTS SSNKVVLNAL LTPGDLVLFD RNNHKSNHHG ALLQAGATPV YLETARNPYG FIGGIDAHCF EESYLRELIA EVAPQRAKEA RPFRLAVIQL GTYDGTIYNA RQVVDKIGHL CDYILFDSAW VGYEQFIPMM ADCSPLLLDL NENDPGILVT QSVHKQQAGF SQTSQIHKKD SHIKGQQRYV PHKRMNNAFM MHASTSPFYP LFAALNINAK MHEGVSGRNM WMDCVVNGIN ARKLILDNCQ HIRPFVPELV DGKPWQSYET AQIAVDLRFF QFVPGEHWHS FEGYAENQYF VDPCKLLLTT PGIDARNGEY EAFGVPATIL ANFLRENGVV PEKCDLNSIL FLLTPAEDMA KLQQLVALLV RFEKLLESDA PLAEVLPSIY KQHEERYAGY TLRQLCQEMH DLYARHNVKQ LQKEMFRKEH FPRVSMNPQE ANYAYLRGEV ELVRLPDAEG RIAAEGALPY PPGVLCVVPG EIWGGAVLRY FSALEEGINL LPGFAPELQG VYIEEHDGRK QVWCYVIKPR DAQSTLLKGE KL
Videos
SDS
Documents