missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Murein hydrolase activator NlpD Protein
A DNA sequence encoding the Escherichia coli (strain K12) Murein hydrolase activator NlpD, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7330-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_417222.1 | |
Murein hydrolase activator NlpD Protein | |
Research Use Only | |
nlpD | |
Recombinant Protein | |
E. coli | |
CSDTSNPPAP VSSVNGNAPA NTNSGMLITP PPKMGTTSTA QQPQIQPVQQ PQIQATQQPQ IQPVQPVAQQ PVQMENGRIV YNRQYGNIPK GSYSGSTYTV KKGDTLFYIA WITGNDFRDL AQRNNIQAPY ALNVGQTLQV GNASGTPITG GNAITQADAA EQGVVIKPAQ NSTVAVASQP TITYSESSGE QSANKMLPNN KPTATTVTAP VTVPTASTTE PTVSSTSTST PISTWRWPTE GKVIETFGAS EGGNKGIDIA GSKGQAIIAT ADGRVVYAGN ALRGYGNLII IKHNDDYLSA YAHNDTMLVR EQQEVKAGQK IATMGSTGTS STRLHFEIRY KGKSVNPLRY LPQR | |
Greater than 90% as determined by SDS-PAGE. |
947011 | |
100 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |