missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Murein hydrolase activator NlpD Protein
A DNA sequence encoding the Escherichia coli (strain K12) Murein hydrolase activator NlpD, was expressed in the hosts and tags indicated.
309.00 € - 3405.00 €
Specifications
Accession Number | NP_417222.1 |
---|---|
Gene ID (Entrez) | 947011 |
Name | Murein hydrolase activator NlpD Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16088240
|
enQuireBio™
QP7330-EC-100UG |
100 μg |
985.00 €
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16008250
|
enQuireBio™
QP7330-EC-1MG |
1 mg |
3405.00 €
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16098240
|
enQuireBio™
QP7330-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16018250
|
enQuireBio™
QP7330-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16028250
|
enQuireBio™
QP7330-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038250
|
enQuireBio™
QP7330-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_417222.1 | |
Murein hydrolase activator NlpD Protein | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
947011 | |
Research Use Only | |
nlpD | |
Recombinant Protein | |
E. coli | |
CSDTSNPPAP VSSVNGNAPA NTNSGMLITP PPKMGTTSTA QQPQIQPVQQ PQIQATQQPQ IQPVQPVAQQ PVQMENGRIV YNRQYGNIPK GSYSGSTYTV KKGDTLFYIA WITGNDFRDL AQRNNIQAPY ALNVGQTLQV GNASGTPITG GNAITQADAA EQGVVIKPAQ NSTVAVASQP TITYSESSGE QSANKMLPNN KPTATTVTAP VTVPTASTTE PTVSSTSTST PISTWRWPTE GKVIETFGAS EGGNKGIDIA GSKGQAIIAT ADGRVVYAGN ALRGYGNLII IKHNDDYLSA YAHNDTMLVR EQQEVKAGQK IATMGSTGTS STRLHFEIRY KGKSVNPLRY LPQR | |
Greater than 90% as determined by SDS-PAGE. |