missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Heat-labile enterotoxin B chain Protein

A DNA sequence encoding the Escherichia coli (strain K12) Heat-labile enterotoxin B chain, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7016-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16087050

Quantity Price
1 3540.93 € / 1mg
3 3306.52 € / 1mg
Estimated Shipment: 17-01-2022

Add to basket


Description

Description

Specifications

Specifications

WP_024167713.1
Greater than 90% as determined by SDS-PAGE.
Research Use Only
eltB
Recombinant Protein
E. coli
APQSITELCS EYHNTQIYTI NDKILSYTES MAGKREMVII TFKSGATFQV EVPGSQHIDS QKKAIERMKD TLRITYLTET KIDKLCVWNN KTPNSIAAIS MEN
Heat-labile enterotoxin B chain Protein
1 mg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
Videos
SDS
Documents
Certificates
Promotions

Promotions