missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli fabF Protein
A DNA sequence encoding the Escherichia coli (strain K12) fabF, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7326-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_415613.1 | |
fabF Protein | |
Research Use Only | |
fabF | |
Recombinant Protein | |
E. coli | |
SKRRVVVTGL GMLSPVGNTV ESTWKALLAG QSGISLIDHF DTSAYATKFA GLVKDFNCED IISRKEQRKM DAFIQYGIVA GVQAMQDSGL EITEENATRI GAAIGSGIGG LGLIEENHTS LMNGGPRKIS PFFVPSTIVN MVAGHLTIMY GLRGPSISIA TACTSGVHNI GHAARIIAYG DADVMVAGGA EKASTPLGVG GFGAARALST RNDNPQAASR PWDKERDGFV LGDGAGMLVL EEYEHAKKRG AKIYAELVGF GMSSDAYHMT SPPENGAGAA LAMANALRDA GIEASQIGYV NAHGTSTPAG DKAEAQAVKT IFGEAASRVL VSSTKSMTGH LLGAAGAVES IYSILALRDQ AVPPTINLDN PDEGCDLDFV PHEARQVSGM EYTLCNSFGF GGTNGSLIFK KI | |
Greater than 90% as determined by SDS-PAGE. |
946665 | |
100 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |